Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205773.2 | NP_991342.1 | 2 - 3 | 277 | 121 | |
GenBank mRNA | AJ132003.2 | - | 2 - 3 | 248 | 121 | |
BC114735.1 | - | 2 - 3 | 277 | 121 | ||
EU744565.1 | - | 2 - 3 | 227 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |