Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAICS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205813.2 | NP_991382.2 | 3 - 4 | 608 | 72 | |
GenBank mRNA | AY344127.1 | - | 4 - 5 | 484 | 72 | |
BC113344.1 | - | 3 - 4 | 608 | 72 | ||
BT030591.2 | - | 3 - 4 | 450 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |