Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025328.2 | NP_001020499.1 | 1 - 2 | 389 | 59 | |
GenBank mRNA | BC118268.1 | - | 1 - 2 | 389 | 59 | |
DQ070851.1 | - | 1 - 2 | 378 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |