Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035080.2 | NP_001030252.2 | 4 - 5 | 865 | 66 | |
GenBank mRNA | AY309439.1 | - | 3 - 4 | 776 | 66 | |
BC123774.1 | - | 4 - 5 | 865 | 66 | ||
BT020946.1 | - | 4 - 5 | 895 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |