Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035080.2 | NP_001030252.2 | 7 - 8 | 1162 | 128 | |
GenBank mRNA | BC123774.1 | - | 7 - 8 | 1162 | 128 | |
BT020946.1 | - | 7 - 8 | 1192 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |