Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075162.2 | NP_001068630.1 | 2 - 3 | 256 | 91 | |
NM_001163274.3 | NP_001156746.2 | 2 - 3 | 256 | 91 | ||
GenBank mRNA | BC120448.1 | - | 2 - 3 | 176 | 91 | |
BE751148.1 | - | 2 - 3 | 256 | 91 | ||
EH159834.1 | - | 2 - 3 | 222 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |