Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075187.1 | NP_001068655.1 | 1 - 2 | 435 | 75 | |
GenBank mRNA | BC119881.1 | - | 1 - 2 | 435 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |