Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075228.2 | NP_001068696.1 | 1 - 2 | 104 | 108 | |
GenBank mRNA | BC114852.1 | - | 1 - 2 | 105 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |