Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075247.1 | NP_001068715.1 | 7 - 8 | 768 | 71 | |
GenBank mRNA | BC119890.1 | - | 7 - 8 | 768 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |