Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075289.1 | NP_001068757.1 | 5 - 6 | 779 | 76 | |
GenBank mRNA | BC123855.1 | - | 5 - 6 | 779 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |