Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075342.1 | NP_001068810.1 | 3 - 4 | 493 | 81 | |
GenBank mRNA | BC120372.1 | - | 3 - 4 | 493 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |