Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075352.2 | NP_001068820.1 | 4 - 5 | 527 | 81 | |
GenBank mRNA | BC120140.1 | - | 4 - 5 | 528 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |