Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075357.1 | NP_001068825.1 | 7 - 8 | 1110 | 90 | |
GenBank mRNA | BC149531.1 | - | 7 - 8 | 1064 | 90 | |
BT026227.1 | - | 7 - 8 | 1110 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |