Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075419.1 | NP_001068887.1 | 8 - 9 | 541 | 67 | |
GenBank mRNA | BC120342.1 | - | 8 - 9 | 541 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |