Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075430.2 | NP_001068898.1 | 5 - 6 | 734 | 108 | |
GenBank mRNA | BC120187.1 | - | 5 - 6 | 736 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |