Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075434.1 | NP_001068902.1 | 2 - 3 | 322 | 78 | |
GenBank mRNA | BC118492.1 | - | 2 - 3 | 322 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |