Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075437.2 | NP_001068905.1 | 7 - 8 | 920 | 76 | |
GenBank mRNA | BC120283.1 | - | 7 - 8 | 921 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |