Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075458.2 | NP_001068926.1 | 3 - 4 | 308 | 88 | |
GenBank mRNA | BC120265.1 | - | 3 - 4 | 309 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |