Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075463.1 | NP_001068931.1 | 5 - 6 | 1240 | 142 | |
GenBank mRNA | BC123859.1 | - | 5 - 6 | 1240 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |