Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNFN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075542.1 | NP_001069010.1 | 2 - 3 | 303 | 96 | |
GenBank mRNA | BC120481.1 | - | 2 - 3 | 303 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |