Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075568.1 | NP_001069036.1 | 6 - 7 | 960 | 60 | |
GenBank mRNA | BC119910.1 | - | 6 - 7 | 960 | 60 | |
BT029851.1 | - | 6 - 7 | 908 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |