Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075579.1 | NP_001069047.1 | 5 - 6 | 886 | 133 | |
GenBank mRNA | BC120049.1 | - | 5 - 6 | 886 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |