Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075625.2 | NP_001069093.1 | 2 - 3 | 213 | 132 | |
GenBank mRNA | BC120045.1 | - | 2 - 3 | 214 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |