Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075647.1 | NP_001069115.1 | 1 - 2 | 40 | 58 | |
GenBank mRNA | BC120431.1 | - | 1 - 2 | 40 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |