Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075647.1 | NP_001069115.1 | 3 - 4 | 210 | 63 | |
GenBank mRNA | BC120431.1 | - | 3 - 4 | 210 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |