Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075663.1 | NP_001069131.1 | 6 - 7 | 1047 | 79 | |
GenBank mRNA | BT026277.1 | - | 6 - 7 | 1047 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |