Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075693.1 | NP_001069161.1 | 7 - 8 | 1012 | 78 | |
GenBank mRNA | BC120300.1 | - | 7 - 8 | 1012 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |