Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075707.1 | NP_001069175.1 | 5 - 6 | 825 | 72 | |
GenBank mRNA | BC119929.1 | - | 5 - 6 | 825 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |