Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075733.1 | NP_001069201.1 | 3 - 4 | 510 | 132 | |
GenBank mRNA | BC119937.1 | - | 3 - 4 | 510 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |