Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075795.2 | NP_001069263.1 | 7 - 8 | 790 | 141 | |
GenBank mRNA | BC122831.1 | - | 7 - 8 | 791 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |