Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075802.1 | NP_001069270.1 | 6 - 7 | 1186 | 66 | |
GenBank mRNA | BC119925.1 | - | 5 - 6 | 1148 | 66 | |
BT026211.1 | - | 6 - 7 | 1186 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |