Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075911.1 | NP_001069379.1 | 6 - 7 | 1573 | 78 | |
GenBank mRNA | BT026269.1 | - | 6 - 7 | 1573 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |