Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075913.1 | NP_001069381.1 | 6 - 7 | 1131 | 105 | |
GenBank mRNA | BC119906.1 | - | 6 - 7 | 1131 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |