Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075924.1 | NP_001069392.1 | 4 - 5 | 991 | 127 | |
GenBank mRNA | BC123495.1 | - | 4 - 5 | 991 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Inventoried Probe spans exons |