Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075940.2 | NP_001069408.1 | 1 - 2 | 412 | 100 | |
GenBank mRNA | BC120182.1 | - | 1 - 2 | 227 | 100 | |
BT025409.1 | - | 1 - 2 | 177 | 100 | ||
BT025454.1 | - | 1 - 2 | 204 | 100 | ||
BT026254.1 | - | 1 - 2 | 218 | 100 | ||
BT026255.1 | - | 1 - 2 | 412 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |