Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075976.2 | NP_001069444.1 | 4 - 5 | 538 | 119 | |
GenBank mRNA | BC120177.1 | - | 4 - 5 | 539 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |