Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076001.2 | NP_001069469.1 | 5 - 6 | 764 | 89 | |
GenBank mRNA | BC120181.1 | - | 5 - 6 | 765 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |