Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076032.2 | NP_001069500.1 | 5 - 6 | 800 | 87 | |
GenBank mRNA | BC120089.1 | - | 5 - 6 | 801 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |