Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2U1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076050.2 | NP_001069518.1 | 1 - 2 | 549 | 64 | |
GenBank mRNA | BC122663.1 | - | 1 - 2 | 550 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |