Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076065.1 | NP_001069533.1 | 8 - 9 | 710 | 83 | |
GenBank mRNA | BC123461.1 | - | 8 - 9 | 710 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |