Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076068.1 | NP_001069536.1 | 15 - 16 | 1822 | 101 | |
GenBank mRNA | BC119838.1 | - | 15 - 16 | 1822 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |