Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SINHCAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076116.1 | NP_001069584.1 | 2 - 3 | 266 | 122 | |
GenBank mRNA | BC119933.1 | - | 2 - 3 | 266 | 122 | |
BC149791.1 | - | 2 - 3 | 249 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |