Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076194.1 | NP_001069662.1 | 5 - 6 | 906 | 75 | |
GenBank mRNA | BT026338.1 | - | 5 - 6 | 906 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |