Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076247.1 | NP_001069715.1 | 3 - 4 | 852 | 122 | |
GenBank mRNA | BC120374.1 | - | 3 - 4 | 852 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |