Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076257.2 | NP_001069725.1 | 7 - 8 | 1424 | 143 | |
GenBank mRNA | BC120276.1 | - | 7 - 8 | 1425 | 143 | |
KP292570.1 | - | 7 - 8 | 1425 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |