Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H9orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076305.2 | NP_001069773.1 | 7 - 8 | 651 | 82 | |
GenBank mRNA | BC122744.1 | - | 7 - 8 | 652 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |