Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076309.1 | NP_001069777.1 | 8 - 9 | 612 | 80 | |
GenBank mRNA | BC120368.1 | - | 8 - 9 | 612 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |