Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076338.1 | NP_001069806.1 | 1 - 2 | 200 | 125 | |
GenBank mRNA | BC120136.1 | - | 1 - 2 | 200 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |