Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076340.1 | NP_001069808.1 | 4 - 5 | 512 | 153 | |
GenBank mRNA | BC120478.1 | - | 4 - 5 | 512 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |