Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076474.2 | NP_001069942.1 | 3 - 4 | 316 | 80 | |
GenBank mRNA | BC122730.1 | - | 3 - 4 | 317 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |